Learn More
Apexbio Technology LLC Betulin 473-98-3 20mg
SDP
Supplier:  Apexbio Technology LLC N129020
Encompass_Preferred
Betulin (CAS 473-98-3) is a naturally occurring pentacyclic triterpene featuring hydroxyl substitutions at the C3 and C28 positions Research indicates that Betulin modulates intracellular signaling pathways in ethanol-exposed hepatic cells in part by attenuating reactive oxygen species (ROS) production and suppressing inflammatory cytokines such as TNF and TNF In studies using human cancer cell lines Betulin has been investigated for its potential to influence cellular responses relevant to pathogenesis These properties support its use as a tool compound in studies of oxidative stress inflammation and carcinogenesis
Catalog No. 50-224-0425