Learn More
Medchemexpress LLC HY-10529 100mg Medchemexpress, Betulinic acid CAS:472-15-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10529100MG
Encompass_Preferred
Medchemexpress, HY-10529 100mg Betulinic acid CAS:472-15-1 Betulinic acid is a natural pentacyclic triterpenoid, acts as a eukaryotic topoisomerase I inhibitor, with an IC50 of 5 μM, and possesses anti-HIV, anti-malarial, anti-inflammatory and anti-tumor properties[3][4]. Betulinic acid acts as a new activator of NF-kB[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8265