Learn More
Cayman Chemical BetulnIc ACD propargyl Ste 5mg
SDP
Supplier:  Cayman Chemical 330305 MG
Encompass_Preferred
An alkyne derivative of betulinic acid; has been used in click chemistry reactions for the synthesis of cytotoxic anticancer betulinic acid derivatives
Catalog No. 50-294-8658