Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bevenopran | 676500-67-7 | MFCD26967979 | 99.8% | 386.44 g/mol | C20H26N4O4 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY10012210MG

Encompass_Preferred

Bevenopran is a peripherally acting μ-opioid receptor antagonist supplied for research use. It has been investigated for the treatment of opioid-induced bowel dysfunction and is provided as a solid or as a DMSO solution for laboratory studies.

  • Peripherally selective μ-opioid receptor antagonist.
  • High purity (>99.8%).
  • White to off-white solid appearance.
  • Available as multiple solid quantities and as a 10 mM solution in DMSO.
  • Molecular weight 386.44 g/mol; formula C20H26N4O4.
  • Recommended storage: powder at -20°C; solution at -80°C for long-term stability.

Catalog No. 50-002-15605


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.