Learn More
Medchemexpress LLC Bexarotene-d4 | 2182068-00-2 | MFCD21363455 | 98.5% | 352.50 g/mol | C24H24D4O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY14171S5MG
Encompass_Preferred
Bexarotene-d4 is a deuterium-labeled analogue of bexarotene, a selective retinoid X receptor (RXR) agonist. Supplied as a purified solid for research use, it is commonly employed as an internal standard or tracer in LC-MS/GC-MS quantitation and metabolism studies involving bexarotene.
  • Deuterium-labeled internal standard for mass spectrometry assays.
  • High isotopic and chemical purity suitable for quantitative analysis.
  • Stable solid form with recommended low-temperature storage.
  • Used in pharmacokinetic, metabolism, and tracer studies.
Catalog No. 50-002-21249