Learn More
Medchemexpress LLC Bezafibrate-d6 | 1219802-74-0 | 98.9% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB0637S1MG
Encompass_Preferred
Bezafibrate-d6 is the deuterium labeled Bezafibrate. Bezafibrate is an agonist of PPAR, with EC50s of 50 μM, 60 μM, and 20 μM for human PPARα, PPARγ, and PPARδ respectively, and 90 μM, 55 μM, and 110 μM for murine PPARα, PPARγ, and PPARδ respectively. It is used as a hypolipidemic agent.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08410