Learn More
Medchemexpress LLC Bezisterim | 1001100-69-1 | 95.0% | 330.46 g·mol⁻¹ | C21H30O3 | 5 MG

Supplier: Medchemexpress LLC HY1080395MG
Bezisterim is an orally active, small-molecule partial NF-κB inhibitor and a synthetic derivative of the endogenous anti-inflammatory steroid β-AET. It is used in preclinical research to study inflammation-related signaling and disease models where modulation of NF-κB activity is relevant.
- Partial NF-κB inhibition useful for inflammation pathway studies.
- Orally active chemical profile suitable for in vivo experiments.
- Molecular formula C21H30O3; molecular weight 330.46 g·mol⁻¹.
- CAS number 1001100-69-1 for unambiguous identification.
- Purity reported as 95.0% for research-grade applications.
- Offered in small solid pack sizes for dosing and formulation flexibility.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.