Learn More
Medchemexpress LLC Bezuclastinib | 1616385-51-3 | 98.8% | C19H17N5O | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1455571MG
Encompass_Preferred
Bezuclastinib (CGT9486) is an orally active, highly selective tyrosine kinase inhibitor that demonstrates potent activity against KIT D816V. It is used in the research of nonadvanced systemic mastocytosis (NonAdvSM).
  • Orally active and highly selective tyrosine kinase inhibitor
  • Potent activity against KIT D816V
  • Used in research of nonadvanced systemic mastocytosis (NonAdvSM)
  • Appears as a white to off-white solid
  • Shows high selectivity to KIT D816V
  • Minimal brain penetrance
  • IC50 of <1 μM against c-Kit and c-kit D816V
Catalog No. 50-003-55870