Learn More
Medchemexpress LLC BGC-20-1531 free base | 736183-35-0 | 98.1% | 492.54 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1984910MM/1ML
Encompass_Preferred
BGC-20-1531 (PGN 1531) free base is a potent and selective prostanoid EP4 receptor antagonist with a pKB of 7.6. It has potential for migraine headache research, competitively antagonizing PGE2-induced vasodilatation in human middle cerebral and meningeal arteries, while showing no effect on responses induced by PGE2 in coronary, pulmonary or renal arteries. In vivo, it causes a dose-dependent antagonism of PGE2-induced increase in canine carotid blood flow.
  • Potent and selective prostanoid EP4 receptor antagonist.
  • Potential for migraine headache research.
  • Competitively antagonizes PGE2-induced vasodilatation of human middle cerebral and meningeal arteries.
  • Shows no effect on responses induced by PGE2 on coronary, pulmonary or renal arteries.
  • Causes a dose-dependent antagonism of PGE2-induced increase in canine carotid blood flow in vivo.
Catalog No. 50-004-00791