Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences (BGE), n-Butyl Glycidyl Ether, (Araldite RD-1) 100 ML
SDP

Supplier:  Electron Microscopy Sciences 12000

Encompass_Preferred

(Butoxymethyl oxirane)
CAS #2426-08-6 C₇H₁₄O₂ F.W. 130.19
Boiling point: 166-168°C

Catalog No. 50-980-272


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.