Learn More
Selleck Chemical LLC BGP-15 2HCl
SDP
Supplier:  Selleck Chemical LLC S83705MG
Encompass_Preferred
BGP-15 is a nicotinic amidoxime derivative with PARP inhibitory activity It has been demonstrated that BGP-15 protects against ischemia-reperfusion injury
Catalog No. 50-000-01115