Learn More
Medchemexpress LLC BGSN3 (Benzyl-Guanine Substrate) | 2996069-91-9 | 98.1% | 381.39 | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HY13916310MM/1ML
Encompass_Preferred
BGSN3 is a click chemistry reagent containing an Azide group, making it suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions. It is also an effective substrate for SNAP-tag and H5 enzymes, with IC50 values of 17.8 μM and 10 μM respectively, and can be used for direct immobilization of these tags on solid surfaces.
  • Good substrate for SNAP-tag and H5 enzymes
  • Contains an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Effective for protein/enzyme labeling using SNAP-tag technology
  • Suitable for direct immobilization on solid surfaces
Catalog No. 50-003-50751