Learn More
AdipoGen BHHT
SDP
Supplier:  AdipoGen CDXB0146M100
Encompass_Preferred
Chemical. CAS 200862-69-7. Formula C30H16F14O4. MW 706.42. Synthetic. Chelating label for europium Eu3. Builds luminescent complexes that can be directly bound to proteins and detected.
Catalog No. 50-256-1655