Learn More
Medchemexpress LLC Bhpi | 56632-39-4 | 98.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1282510MG
Encompass_Preferred
BHPI is a nuclear receptor ERα inhibitor that activates PLCγ and UPR. It exhibits potent antitumor activity against various cancers, including breast, endometrial, and ovarian cancers.
  • Inhibits proliferation of ERα-positive breast, endometrial, and ovarian cancer cells.
  • Inhibits cell proliferation in two-dimensional adherent cultures of T47D, ERαY537S-4, and ERαD538G-1 cells.
  • Demonstrates in vivo tumor growth inhibition and regression in mouse xenograft models using MCF-7 tumors.
Catalog No. 50-003-42877