Learn More
Medchemexpress LLC BHQ-2 N-hydroxysuccinimide ester | 916753-62-3 | MFCD28160399 | 97.7% | 603.58 g/mol | C29H29N7O8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY4298410MG
Encompass_Preferred
BHQ-2 NHS is a non-emitting quencher containing a polyaromatic-azo backbone with a terminal N-hydroxysuccinimide (NHS) ester. It is designed for covalent labeling of primary amines on proteins and amine-modified oligonucleotides, and for use as an acceptor in fluorescence resonance energy transfer (FRET) and other fluorescence-based assays.
  • Provides strong quenching across 560-670 nm.
  • NHS ester reacts with primary amines for covalent labeling.
  • High purity, approximately 97.7%.
  • Molecular weight 603.58 g/mol.
  • Suitable for oligonucleotide and protein labeling in FRET assays.
Catalog No. 50-002-98600