Learn More
Selleck Chemical LLC BI-2493-E1564-1MG
SDP
Supplier:  Selleck Chemical LLC E15641MG
Encompass_Preferred
BI-2493 a structural analogue of BI-2865 is a highly selective inhibitor of pan-KRAS BI-2493 attenuates tumor growth and can be used for cancer research
Catalog No. 50-313-7212