Learn More
Medchemexpress LLC BI 2536 | 755038-02-9 | 99.9% | 500 MG
SDP
Supplier:  Medchemexpress LLC HY50698500MG
Encompass_Preferred
BI 2536 is a dual inhibitor of PLK1 and BRD4, demonstrating IC50 values of 0.83 nM and 25 nM, respectively. This compound also suppresses IFNB (encoding IFN-β) gene transcription and is intended for research use only.
  • Dual PLK1 and BRD4 inhibitor
  • Suppresses IFNB gene transcription
  • Suitable for reconstitution in DMSO at 10 mM
  • Available for research use only
Catalog No. 50-003-58750