Learn More
Selleck Chemical LLC BI-3406 5mg
SDP
Supplier:  Selleck Chemical LLC S89165MG
Encompass_Preferred
BI-3406 (compound I-13) is a potent, selective and orally active inhibitor of the interaction betweenKRAS andSon of Sevenless 1 (SOS1) with IC50 of 5 nM. BI-3406 reduces the formation of GTP-loaded KRAS, and inhibits MAPK pathway signaling. BI-3406 exhibits anti-tumor activities. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3433
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.