Learn More
Medchemexpress LLC HY-108705 10mg Medchemexpress, BI-3802 CAS:2166387-65-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10870510MG
Encompass_Preferred
Medchemexpress, HY-108705 10mg BI-3802 CAS:2166387-65-9 BI-3802 is a highly potent BCL6 degrader, inhibiting the BTB domain of BCL6, with an IC50 of ≤3 nM; BI-3802 has antitumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8616