Learn More
Selleck Chemical LLC BI-4020 S8921-1MG
SDP
Supplier:  Selleck Chemical LLC S89211MG
Encompass_Preferred
BI-4020 is an orally active non-covalent EGFR inhibitor with IC50 of 0 6 nM and 0 2 nM for EGFRdel19 T790M C797S as biomarker potency and anti-proliferation potency in BaF3 cells respectively
Catalog No. 50-217-3436