Learn More
Medchemexpress LLC BI-4020 | 2664214-60-0 | 98.82% | 50 MG
SDP
Supplier:  Medchemexpress LLC HY12955050MG
Encompass_Preferred
BI-4020 is a fourth-generation, orally active, and non-covalent EGFR tyrosine kinase inhibitor. It exhibits high kinome selectivity and favorable DMPK properties. This compound is stable under recommended storage conditions.
  • Inhibits the triple mutant EGFR del19 T790M C797S variant (IC50=0.2 nM in BaF3 cell lines).
  • Inhibits the double mutant EGFR del19 T790M and primary mutant EGFR del19 (IC50=1 nM).
  • Shows activity against EGFR wt (IC50=190 nM).
  • For research use only by suitably qualified experienced scientists.
Catalog No. 50-003-42352