Learn More
Medchemexpress LLC BI-6015 | 93987-29-2 | 99.9% | 331.35 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY108469
Encompass_Preferred
BI-6015 is an antagonist of hepatocyte nuclear factor 4α (HNF4α) that can inhibit the expression of known HNF4α target genes. It represses insulin promoter activity through HNF4α antagonism. BI-6015 can be used for the research of cancer and diabetes.
  • HNF4α antagonist
  • Inhibits expression of HNF4α target genes
  • Represses insulin promoter activity
  • Applicable for cancer research
  • Applicable for diabetes research
Catalog No. 50-003-47749