Learn More
Medchemexpress LLC HY-111925 5mg Medchemexpress, BI-749327 CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1119255MG
Encompass_Preferred
Medchemexpress, HY-111925 5mg BI-749327 CAS: BI-749327 is a potent, high selectivity and orally bioavailable TRPC6 antagonist, with IC 50 s of 13 nM, 19 nM and 15 nM for mouse, human and guinea pig TRPC6, respectively. BI-749327 is 85-fold more selective for mouse TRPC6 than TRPC3 and 42-fold versus TRPC7 [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2408