Learn More
TARGETMOL CHEMICALS INC BI-78D3 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T678425MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BI-78D3, a competitive JNK inhibitor. IC50 of BI-78D3 is 280 nM that displays > 100 fold selectivity over p38alpha, and have no activity at mTOR and PI-3K. Purity 97.89%
Catalog No. 50-287-9096