Learn More
Selleck Chemical LLC BI-847325 5mg 1207293-36-4
SDP
Supplier:  Selleck Chemical LLC S78435MG
Encompass_Preferred
BI-847325 is an orally bioavailable, and selective dual MEK/Aurora kinase inhibitor with IC50 of 3 nM, 25 nM, 15 nM, 25 nM, and 4 nM for Xenopus laevis Aurora B, human Aurora A and Aurora C, as well as human MEK1 and MEK2, respectively. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5329