Learn More
Alkali Scientific Bi-directional Liquid Transfer
SDP
Supplier:  Alkali Scientific EFC721
Encompass_Preferred
The 5L High Efficiency Erlenmeyer Flask comes with a Bi-directional Transfer Cap featuring a TPE Tube (50cm length 1/4in ID 3/8in OD) for seamless and precise liquid transfers The flask also includes a Vent Filter (0 22 m 24mm) to maintain a sterile environment crucial for sensitive samples Each pack contains 1 flask and each case contains 4 packs providing ample supply for laboratory needs This Erlenmeyer flask is designed to ensure high efficiency in various lab applications requiring a volume capacity of 5 liters The sterile nature of the flask eliminates contamination concerns safeguarding the integrity of your experiments 5L High Efficiency Erlenmeyer Flask 1 system per double wrapped sterile bag - 4 systems per case Flask made of USP grade VI virgin polycarbonate Untreated super smooth interior surface ideal for suspended cultures Attached bi-directional transfer cap Attached thermoplastic elastomer (TPE) external transfer tube No attachment hardware included tubing come
Catalog No. 50-302-8574