Learn More
Medchemexpress LLC Bi-mc-vc-pab-mmae | 1620837-70-8 | 98.4% | 1413.66 | C71H104N12O18 | 5 MG

Supplier: Medchemexpress LLC HY1418335MG
Bi-Mc-VC-PAB-MMAE is a drug-linker conjugate used in antibody-drug conjugate (ADC) research. It links a Bi-Mc-Val-Cit-PAB cleavable linker to the tubulin inhibitor monomethyl auristatin E (MMAE) and is supplied for preclinical ADC construction and linker evaluation. Reported purity is 98.43%; molecular formula C71H104N12O18; molecular weight 1413.66 g/mol. Store at -20°C, protected from light, under nitrogen.
- Drug-linker conjugate for ADC research
- Cleavable Val-Cit-PAB linker coupled to MMAE
- Purity 98.4% as reported
- Molecular formula C71H104N12O18; molecular weight 1413.66 g/mol
- Store at -20°C, protected from light, under nitrogen
- Supplied as a solid in small research quantities (5 MG shown)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.