Learn More
Medchemexpress LLC Bi2536-peg2-halo | 99.4% | 790.39 g/mol | C39H60ClN7O8 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY15949310MG
Encompass_Preferred
A bifunctional chemical probe that links a PLK1 inhibitor to a Halo-tag ligand via a PEG2 spacer, designed for cellular target engagement and imaging studies.
  • Combines PLK1 inhibitory activity with Halo-tag targeting.
  • High stated purity (99.44%).
  • Soluble in DMSO at 100 mg/mL for preparing concentrated stocks.
  • Suitable for cellular target validation and imaging of Halo-tag fusion proteins.
  • Available in small research pack sizes, including 10 mg, for screening and probe studies.
Catalog No. 50-002-87954