Learn More
Medchemexpress LLC BI605906 | 960293-88-3 | 99.7% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1301910MM/1ML
Encompass_Preferred
BI605906 is a chemical probe and a novel IKKβ inhibitor, demonstrating improved selectivity. It inhibits IKKβ with an IC50 value of 380 nM when assayed at 0.1 mM ATP and is intended for research use only.
  • Targets IKKβ as a selective inhibitor.
  • Inhibits IKKβ with an IC50 value of 380 nM at 0.1 mM ATP.
  • Shows high selectivity among over 100 tested protein kinases.
  • Partially inhibits IL-1-stimulated activation of IKKε/TBK1.
  • Suppresses TNFα-induced degradation of NF-KB negative regulator IκB in primary mouse hepatocytes.
  • Strongly inhibits TNF-α-dependent expression of various inflammatory mediators.
  • Does not suppress mTOR signaling or activate AMPK.
Catalog No. 50-003-40999