Learn More
Medchemexpress LLC Bi605906 | 960293-88-3 | 99.7% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13019100MG
Encompass_Preferred
BI605906 is a chemical probe that functions as a novel IKKβ inhibitor. It demonstrates an IC50 value of 380 nM when assayed at 0.1 mM ATP. This compound was originally designed by Boehringer Ingelheim.
  • Novel IKKβ inhibitor
  • IC50 of 380 nM at 0.1 mM ATP
  • Exhibits improved selectivity
  • Partially inhibits IL-1-stimulated activation of IKKε/TBK1
Catalog No. 50-003-40998