Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bibf 1202 | 894783-71-2 | MFCD28167767 | 99.8% | 525.60 g/mol | C30H31N5O4 | 10 MG
SDP

Supplier:  Medchemexpress LLC HY1599210MG

Encompass_Preferred

BIBF 1202 is the carboxylate metabolite of nintedanib and a VEGFR2 kinase inhibitor (reported IC50 ≈ 62 nM). Supplied as a powder for research use, it is intended for kinase inhibition assays, metabolite and ADME studies, and target validation.

  • Carboxylate metabolite of nintedanib with VEGFR2 inhibitory activity (IC50 ≈ 62 nM).
  • High purity (reported 99.82%).
  • Molecular weight 525.60 g/mol; formula C30H31N5O4.
  • Soluble in DMSO at ≥ 45 mg/mL.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years).
  • Supplied in small research quantities suitable for in vitro and in vivo studies.

Catalog No. 50-002-85474


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.