Learn More
TARGETMOL CHEMICALS INC BIBO3304 TFA 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T105395MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BIBO3304 TFA is an effective orally active and selective neuropeptide Y (NPY) Y1 receptor antagonist (IC500.38 and 0.72 nM for the human and the rat Y1 receptor). purity: 98%
Catalog No. 50-258-1710