Learn More
Selleck Chemical LLC Dabigatran Etexilate 50mg 211915-06-9 BIBR-1048
SDP
Supplier:  Selleck Chemical LLC S215450MG
Encompass_Preferred
Dabigatran Etexilate (BIBR-1048) is the prodrug of dabigatran, a potent, nonpeptidic small molecule that specifically and reversibly inhibits both free and clot-bound thrombin by binding to the active site of the thrombin molecule. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-524-6