Learn More
Medchemexpress LLC HY-17353 10mg Medchemexpress, BIBR 1532 CAS:321674-73-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1735310MG
Encompass_Preferred
Medchemexpress, HY-17353 10mg BIBR 1532 CAS:321674-73-1 BIBR 1532 is a potent, selective and non-competitive telomerase inhibitor with IC50 of 100 nM in a cell-free assay. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1935