Learn More
Selleck Chemical LLC BIBR 1532 S1186-1g
SDP
Supplier:  Selleck Chemical LLC S11861G
Encompass_Preferred
BIBR 1532 is a potent selective non-competitive telomerase inhibitor with IC50 of 100 nM in a cell-free assay No inhibition of DNA and RNA polymerases including HIV reverse transcriptase are observed at concentrations vastly exceeding the IC50 for telomerase BIBR 1532 induces apoptosis in cancer cells
Catalog No. 50-311-1188