Learn More
Being Scientific BIC-60C - 2.4 cu ft (68 liters),14 - 176 ℉ , -10 - 80℃
Supplier:  Being Scientific BLC15060U
Encompass_Preferred
BEING Cooling Incubators allow users to carry out applications that require temperatures ranging from -10° to +80°C. These incubators are ideal for biochemical oxygen demand (BOD) testing, bacterial research, seed germination, fish/insect breeding and much more. The BEING Cooling Incubator provides accurate and dependable temperature control. Each cooling incubator uses environmentally friendly refrigerants that ensure rapid cooling does not come at the expense of the environment.
Catalog No. 50-002-17579