Learn More
Bioworld Bicarbonate Buffer, 1.0M, pH 8.5, 500 mL
Supplier:  Bioworld 402200073
Encompass_Preferred
Product DescriptionBicarbonate buffer has a concentration of 1.0M and a pH at 8.5.Bicarbonate buffering system is important in different cellular processes such as its use to maintain the pH level of mammalian blood, the formation of acid in the lumen of the stomach, and to neutralize the pH of the chyme.For laboratory use only. Not fit for agricultural, household, clinical use or human consumption.
Catalog No. 50-255-436