Learn More
DIASYS DIAGNOSTIC SYSTEMS USA LLC DiaSys Bicarbonate Standard FS is an aqueous standard (6x3mL)
Supplier:  DIASYS DIAGNOSTIC SYSTEMS USA LLC 109509911030
Encompass_Preferred
DiaSys Bicarbonate Standard is an aqueous standard for use in tests for quantitative in vitro determination of bicarbonate/total CO2. For veterinary or research use only. The standard is used to calibrate Bicarbonate assays. DiaSys Bicarbonate Standard, both opened and unopened, must be stored refrigerated. Avoid contamination and protect from light. Once opened, DiaSys Bicarbonate Standard is stable for 3 months if kept refrigerated and recapped immediatley after use.
Catalog No. NC2874327