Learn More
Bioworld Bicinchoninic Acid, Disodium Salt Hydrate, 10 g
Supplier:  Bioworld 402000911
Encompass_Preferred
A reagent used for absorbance based measurement of protein.Technical Specifications:Also known as 4,4'-Dicarboxy-2,2'-Biquinoline; BCA; 2,2'-Biquinoline-4,4'-Dicarboxylate; 2,2'-Biquinoline-4,4'-Dicarboxylic acidAppearance: Colorless to pale yellow powder
Catalog No. 50-255-400