Learn More
Selleck Chemical LLC Bictegravir 5mg 1611493-60-7 GS-9883
SDP
Supplier:  Selleck Chemical LLC S59115MG
Encompass_Preferred
Bictegravir (GS-9883) is a novel, potent, once-daily, unboosted inhibitor of HIV-1 integrase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8311