Learn More
Medchemexpress LLC Bicuculline (methochloride) | 38641-83-7 | 99.8% | 417.84 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13779050MG
Encompass_Preferred
Bicuculline methochloride is a selective GABAA receptor antagonist with an IC50 value of 3 μM. It induces clonic tonic convulsions in mammals and blocks Ca2+ activated potassium channels, making it useful in studies of epilepsy and related psychiatric disorders.
  • Selective GABAA receptor antagonist (IC50 3 μM)
  • Induces clonic tonic convulsions in mammals
  • Blocks Ca2+ activated potassium channels (SK1 and SK2)
  • Used in epilepsy and related psychiatric disorder research
  • Creates animal epilepsy models by inhibiting inhibitory neurotransmission
  • Increases capillary blood flow in the epileptic focus
Catalog No. 50-003-49518