Learn More
Medchemexpress LLC Bicyclo[3.1.1]heptane-2-ethanol, 6,6-dimethyl- | 4747-61-9 | 98.1% | 168.28 | 500 G
SDP
Supplier:  Medchemexpress LLC 500GHYZ16070
Encompass_Preferred
Hydronopol is a saturated alcohol terpene derivative. It exhibits central inhibitory, anticonvulsant, analgesic, anticholinergic, and antihypertensive activities, making it suitable for research in neurological and cardiovascular diseases.
  • Saturated alcohol terpene derivative
  • Exhibits central inhibitory, anticonvulsant, analgesic, anticholinergic, and antihypertensive activities
  • Can be used for research in neurological and cardiovascular diseases
Catalog No. 50-003-65812