Learn More
Cayman Chemical BIfeprunoxmesylate 10mg
SDP
Supplier:  Cayman Chemical 2952310 MG
Encompass_Preferred
An atypical antipsychotic; dopamine D2 receptor partial agonist and an agonist of the serotonin (5-HT) receptor subtype 5-HT1A (Kis = 2.2 and 9.3 nM, respectively); inhibits apomorphine-induced climbing behavior in mice (ED50 = 0.1 mg/kg) and the conditioned avoidance response in rats (ED50 = 0.8 mg/kg); decreases basal prepulse inhibition of the acoustic startle response in rats by 42% at 10 mg/kg
Catalog No. 50-298-9351