Learn More
Electron Microscopy Sciences Big-Tip Cryo-Markers™, Yellow
SDP
Supplier:  Electron Microscopy Sciences 6205503Y
Encompass_Preferred
Easy-to-use high quality cryogenic permanent ink markers that will mark most surfaces and dries in 5-7 minutes. They feature a thermal resistance of ink: From -196 deg C to +200 deg C. Solid ink and fade and water proof.
Color: Yellow
Catalog No. 50-193-0735