Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Rubbermaid Big Wheel Cart Axle, 33.38 in., 0.5 in.

Supplier:  MSC 71425953

Encompass

This replacement axle with hardware is designed for Rubbermaid Commercial Products 7.5 cubic foot Big Wheel Carts, specifically models FG564200BLA and FG564261BLA. It includes one axle, two cotter pins, and two washers for a complete installation.

  • Designed for 7.5 cubic foot Big Wheel Carts
  • Includes one axle, two cotter pins, and two washers
  • Length: 33.38 in.
  • Diameter: 0.5 in.
  • Weight: 4 lbs

Catalog No. 50-283-5840


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.