Learn More
ATCC Big-Six Escherichia coli Genomic DNA Panel (MP-10) - six non-O157 STEC genomic DNA preparations
Supplier:  ATCC MP10
Encompass
Big-Six Escherichia coli Genomic DNA Panel (MP-10) is a quality-control panel containing purified genomic DNA preparations from the six major non-O157 Shiga toxin-producing Escherichia coli (STEC) serogroups. The panel is intended for assay development, validation, and food-testing quality-control workflows.
  • Contains purified genomic DNA from serogroups O26, O45, O103, O111, O121, and O145.
  • Provides material for assay development, validation, and quality control.
  • Components include genomic DNA preparations derived from representative strains of each serogroup.
  • For laboratory research use only; not for human or diagnostic use.
  • Component genomic DNAs are listed at biosafety level 1 on their individual product pages.
Catalog No. NC3591525