Learn More
Selleck Chemical LLC BIIB059-A2598-1MG
SDP
Supplier:  Selleck Chemical LLC A25981MG
Encompass_Preferred
BIIB059 is a humanized IgG1- antibody targeting to C-type lectin domain family 4 member C (CLEC4C) It can be used for cutaneous lupus erythematosus (CLE) study MW 146 34 KD
Catalog No. 50-313-5249