Learn More
Medchemexpress LLC BIIB091 | 2247614-80-6 | 99.9% | 542.64 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13998450MG
Encompass_Preferred
BIIB091 is a potent, selective, orally active, and reversible BTK inhibitor, with an IC50 of <0.5 nM. It binds the BTK protein to sequester TYR-551 into an inactive conformation with excellent affinity. BIIB091 can be used for the research of multiple sclerosis.
  • Potent and selective BTK inhibitor
  • Orally active and reversible
  • Excellent affinity
  • Used for multiple sclerosis research
Catalog No. 50-003-47972