Learn More
Selleck Chemical LLC BIIB129 E4643-5MG
SDP
Supplier:  Selleck Chemical LLC E46435MG
Encompass_Preferred
BIIB129 is a unique brain-penetrant covalent inhibitor of Bruton s tyrosine kinase (BTK) with high kinome selectivity It exhibits efficacy in targeting B-cell proliferation in the central nervous system (CNS) making it a promising immunomodulatory therapy for multiple sclerosis (MS)
Catalog No. 50-000-39332