Learn More
SEKISUI DIAGNOSTICS LLC Total Bile Acids Assay Kit
Supplier:  SEKISUI DIAGNOSTICS LLC DZ042AK01
Encompass_Preferred
Total Bile Acids Assay Kit is intended for the in vitro quantitative determination of total bile acids (TBA) in human serum. No longer includes the Bile Acid Calibrator. The part number for the calibrator is NC1777599.
Catalog No. NC9846554